Product Details

SNP ID
rs8008540
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:74481477 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTACAACAAGGCAAATGGACTAA[C/T]ACATGAAGGCTTACATGAATAATTT
Phenotype
MIM: 601015 MIM: 609999
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR4709 PubMed Links

Gene Details

Gene
MIR4709
Gene Name
microRNA 4709
There are no transcripts associated with this gene.

Gene
NPC2
Gene Name
NPC intracellular cholesterol transporter 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006432.3 Intron NP_006423.1
XM_017020930.1 Intron XP_016876419.1
Gene
SYNDIG1L
Gene Name
synapse differentiation inducing 1 like
There are no transcripts associated with this gene.

View Full Product Details