Product Details

SNP ID
rs7477
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:16342702 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGCAGAGATCAAGATGACCCTAGTC[A/C]ACGGAACCAGCAGCCCAGGTCAGCC
Phenotype
MIM: 608139 MIM: 605947
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CENPV PubMed Links

Gene Details

Gene
CENPV
Gene Name
centromere protein V
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181716.2 996 UTR 3 NP_859067.2
Gene
PIGL
Gene Name
phosphatidylinositol glycan anchor biosynthesis class L
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004278.3 996 Intron NP_004269.1
XM_011524080.2 996 Intron XP_011522382.1
XM_017025349.1 996 Intron XP_016880838.1
XM_017025350.1 996 Intron XP_016880839.1
XM_017025351.1 996 Intron XP_016880840.1
XM_017025352.1 996 UTR 3 XP_016880841.1
XM_017025353.1 996 Intron XP_016880842.1
XM_017025354.1 996 UTR 3 XP_016880843.1
XM_017025355.1 996 Intron XP_016880844.1
XM_017025356.1 996 UTR 3 XP_016880845.1

View Full Product Details