Product Details

SNP ID
rs498778
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52419877 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCAGAGGGGCCAGTCCAACAGAGG[C/T]TTCCATGGATTTCTGCCTGAAGACA
Phenotype
MIM: 603089
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BAP1 PubMed Links

Gene Details

Gene
BAP1
Gene Name
BRCA1 associated protein 1
There are no transcripts associated with this gene.

Gene
PHF7
Gene Name
PHD finger protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278221.2 990 Silent Mutation GGC,GGT G77G NP_001265150.1
NM_001321126.1 990 Silent Mutation GGC,GGT G77G NP_001308055.1
NM_001321127.1 990 Silent Mutation GGC,GGT G77G NP_001308056.1
NM_016483.6 990 Silent Mutation GGC,GGT G77G NP_057567.3
XM_011533826.2 990 Silent Mutation GGC,GGT G77G XP_011532128.1

View Full Product Details