Product Details

SNP ID
rs4801160
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56407292 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGTCTCCAAAAGCCTAGTTATC[G/T]TTTTTTCCACAGCCAGCATTTTTAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF582-AS1 PubMed Links

Gene Details

Gene
ZNF582-AS1
Gene Name
ZNF582 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
ZNF583
Gene Name
zinc finger protein 583
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159860.1 Intron NP_001153332.1
NM_001159861.1 Intron NP_001153333.1
NM_152478.2 Intron NP_689691.2
XM_005258558.1 Intron XP_005258615.1
XM_011526517.2 Intron XP_011524819.1
XM_017026346.1 Intron XP_016881835.1
XM_017026347.1 Intron XP_016881836.1
XM_017026348.1 Intron XP_016881837.1
XM_017026349.1 Intron XP_016881838.1
XM_017026350.1 Intron XP_016881839.1
XM_017026351.1 Intron XP_016881840.1
XM_017026352.1 Intron XP_016881841.1
XM_017026353.1 Intron XP_016881842.1

View Full Product Details