Product Details

SNP ID
rs10840686
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:8109054 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTTGTAGGAATTTGCTCAGACTCCT[C/G]GTGGCAGGGGAAGCATTTGTTTTTT
Phenotype
MIM: 605306
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CLEC4A PubMed Links

Gene Details

Gene
CLEC4A
Gene Name
C-type lectin domain family 4 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016184.3 Intron NP_057268.1
NM_194447.2 Intron NP_919429.2
NM_194448.2 Intron NP_919430.1
NM_194450.2 Intron NP_919432.1
XM_011520684.2 Intron XP_011518986.1
XM_017019382.1 Intron XP_016874871.1

View Full Product Details