Product Details

SNP ID
rs4806980
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4347172 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTTTTTTCTGTTTTTGCTGTTAT[C/G]GCCTTCAACTGATTGGATGAGGCCC
Phenotype
MIM: 607881
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MPND PubMed Links

Gene Details

Gene
MPND
Gene Name
MPN domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159846.2 Intron NP_001153318.1
NM_001300862.1 Intron NP_001287791.1
NM_032868.5 Intron NP_116257.2
XM_006722926.2 Intron XP_006722989.1
Gene
STAP2
Gene Name
signal transducing adaptor family member 2
There are no transcripts associated with this gene.

View Full Product Details