Product Details

SNP ID
rs3755340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:70902533 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAATCCCACACATTCCCAGGGGA[A/G]GATCTAAAGTCACTGCCTGTGACAC
Phenotype
MIM: 604295
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VAX2 PubMed Links

Gene Details

Gene
VAX2
Gene Name
ventral anterior homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012476.2 Intron NP_036608.1
XM_006711982.3 Intron XP_006712045.1
XM_011532750.2 Intron XP_011531052.1
XM_011532751.2 Intron XP_011531053.1

View Full Product Details