Product Details

SNP ID
rs35389602
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:65060598 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAAATGAATAAAAATAAAAACTG[G/T]CAGTATAGTAAAAGCAGTGGTCTGT
Phenotype
MIM: 616889
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CEP68 PubMed Links

Gene Details

Gene
CEP68
Gene Name
centrosomal protein 68
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319100.1 Intron NP_001306029.1
NM_001319101.1 Intron NP_001306030.1
NM_015147.2 Intron NP_055962.2
XM_005264219.3 Intron XP_005264276.1
XM_017003633.1 Intron XP_016859122.1

View Full Product Details