Product Details

SNP ID
rs13180322
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:175484230 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTGATGTTCAAGACACCTCTGTAA[C/T]GAGATGTTTGATCTTTACCCAGTTT
Phenotype
MIM: 615569
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SFXN1 PubMed Links

Gene Details

Gene
SFXN1
Gene Name
sideroflexin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322977.1 Intron NP_001309906.1
NM_001322978.1 Intron NP_001309907.1
NM_001322980.1 Intron NP_001309909.1
NM_001322981.1 Intron NP_001309910.1
NM_001322982.1 Intron NP_001309911.1
NM_001322983.1 Intron NP_001309912.1
NM_022754.6 Intron NP_073591.2

View Full Product Details