Product Details

SNP ID
rs13028484
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:42508032 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGATTACCTCTTTTTCCTGAGGCCC[A/G]TTCTTGCTGGTTTCTGCATCCCCAT
Phenotype
MIM: 609050
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MTA3 PubMed Links

Gene Details

Gene
MTA3
Gene Name
metastasis associated 1 family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282755.1 Intron NP_001269684.1
NM_001282756.1 Intron NP_001269685.1
NM_020744.3 Intron NP_065795.1
XM_005264456.2 Intron XP_005264513.1
XM_005264458.2 Intron XP_005264515.1
XM_005264459.2 Intron XP_005264516.1
XM_017004561.1 Intron XP_016860050.1
XM_017004562.1 Intron XP_016860051.1

View Full Product Details