Product Details

SNP ID
rs3796075
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:42495270 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AACTGAAAATGTGAGTGCTGTCTTT[A/T]AAGTAAAATCACTCTCGGTGGCTTC
Phenotype
MIM: 606767 MIM: 609050
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KCNG3 PubMed Links

Gene Details

Gene
KCNG3
Gene Name
potassium voltage-gated channel modifier subfamily G member 3
There are no transcripts associated with this gene.

Gene
MTA3
Gene Name
metastasis associated 1 family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282755.1 Intron NP_001269684.1
NM_001282756.1 Intron NP_001269685.1
NM_020744.3 Intron NP_065795.1
XM_005264456.2 Intron XP_005264513.1
XM_005264458.2 Intron XP_005264515.1
XM_005264459.2 Intron XP_005264516.1
XM_017004561.1 Intron XP_016860050.1
XM_017004562.1 Intron XP_016860051.1

View Full Product Details