Product Details

SNP ID
rs12402234
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:145891531 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAATATAAGTTACCCAGTTGTCT[G/A]TGAGAATCCAACTGCCTTTACTGTC
Phenotype
MIM: 604042
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
ANKRD35 PubMed Links

Gene Details

Gene
ANKRD35
Gene Name
ankyrin repeat domain 35
There are no transcripts associated with this gene.

Gene
ITGA10
Gene Name
integrin subunit alpha 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303040.1 4353 UTR 3 NP_001289969.1
NM_001303041.1 4353 UTR 3 NP_001289970.1
NM_003637.4 4353 UTR 3 NP_003628.2
XM_005277436.3 4353 UTR 3 XP_005277493.1
XM_011510083.2 4353 UTR 3 XP_011508385.1
XM_011510084.1 4353 UTR 3 XP_011508386.1
XM_017002622.1 4353 UTR 3 XP_016858111.1
XM_017002623.1 4353 UTR 3 XP_016858112.1
XM_017002624.1 4353 UTR 3 XP_016858113.1
XM_017002625.1 4353 UTR 3 XP_016858114.1
XM_017002626.1 4353 UTR 3 XP_016858115.1
XM_017002627.1 4353 Intron XP_016858116.1
XM_017002628.1 4353 UTR 3 XP_016858117.1

View Full Product Details