Product Details
- SNP ID
-
rs334878
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:55224050 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCATCCTCATACCAGTGTGGGAAGC[A/G]GCAGGGGCCTAACTGATTCCTCTTT
- Phenotype
-
MIM: 607752
MIM: 614086
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CCNO
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116834645] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCNO
- Gene Name
- cyclin O
There are no transcripts associated with this gene.
- Gene
- MCIDAS
- Gene Name
- multiciliate differentiation and DNA synthesis associated cell cycle protein
View Full Product Details