Product Details

SNP ID
rs4759455
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:130341333 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TGAAAGACTGGTACCACCCTACCTT[C/G]CAATTCCTAATTCTCTGTTTCAGCT
Phenotype
MIM: 605571
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PIWIL1 PubMed Links

Gene Details

Gene
PIWIL1
Gene Name
piwi like RNA-mediated gene silencing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190971.1 Intron NP_001177900.1
NM_004764.4 Intron NP_004755.2
XM_011539002.2 Intron XP_011537304.1
XM_011539003.2 Intron XP_011537305.1
XM_011539004.2 Intron XP_011537306.1
XM_011539005.1 Intron XP_011537307.1
XM_011539006.2 Intron XP_011537308.1
XM_017020229.1 Intron XP_016875718.1

View Full Product Details