Product Details
- SNP ID
-
rs12522073
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:179060451 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTCCTGGCTTGGGCCATCCAGGCT[C/T]GGCTAGGATTGCCGGCCTGGGGTTG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF354C
PubMed Links
Gene Details
- Gene
- ZNF354C
- Gene Name
- zinc finger protein 354C
View Full Product Details