Product Details

SNP ID
rs877068
Assay Type
Validated
NCBI dbSNP Submissions
49
Location
Chr.1:110155423 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAATATGAATTACTGTATACACTTA[C/T]TCTCATGTTCAGAACCAGGTATGTT
Phenotype
MIM: 610299
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC6A17 PubMed Links

Gene Details

Gene
SLC6A17
Gene Name
solute carrier family 6 member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010898.2 Intron NP_001010898.1
XM_006710643.2 Intron XP_006710706.1

View Full Product Details