Product Details

SNP ID
rs304485
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:89400153 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTCTATACATCTGTCTTTATACCA[A/T]TGTTGTAATATTGTGATTACTGTAG
Phenotype
MIM: 147690
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
IFIT1 PubMed Links

Gene Details

Gene
IFIT1
Gene Name
interferon induced protein with tetratricopeptide repeats 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270927.1 Intron NP_001257856.1
NM_001270928.1 Intron NP_001257857.1
NM_001270929.1 Intron NP_001257858.1
NM_001270930.1 Intron NP_001257859.1
NM_001548.4 Intron NP_001539.3

View Full Product Details