Product Details
- SNP ID
-
rs1224329
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:37212461 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CATTCCCAGACGGACAGGTAGTCAT[A/G]AGTTCACTGCAATGGTGGAGCAGAG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM217
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74359974] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TMEM217
- Gene Name
- transmembrane protein 217
View Full Product Details