Product Details

SNP ID
rs1360438
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.13:96885610 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTCCTGAGGGTTGGGCATGATGTCA[A/G]TGTTCAGCTCTTTACCACTGTCCAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSP90AB6P PubMed Links

Gene Details

Gene
HSP90AB6P
Gene Name
heat shock protein 90 alpha family class B member 6, pseudogene
There are no transcripts associated with this gene.

View Full Product Details