Product Details

SNP ID
rs8192793
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41193981 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGATGGGTGGAGCTCAGTTGGGA[C/G]TTCCTGGGACTGGGGAGGAAGGAGA
Phenotype
MIM: 611529
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CYP2S1 PubMed Links

Gene Details

Gene
CYP2S1
Gene Name
cytochrome P450 family 2 subfamily S member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030622.7 Intron NP_085125.1

View Full Product Details