Product Details

SNP ID
rs17468739
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:89306192 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGAATGCTTACGTAAAGCTGAAG[A/C]GTTAATCCAGCAAGAGCATGCTGAC
Phenotype
MIM: 147040
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IFIT2 PubMed Links

Gene Details

Gene
IFIT2
Gene Name
interferon induced protein with tetratricopeptide repeats 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001547.4 411 Missense Mutation GAG,GCG E79A NP_001538.4
Gene
LOC105378419
Gene Name
uncharacterized LOC105378419
There are no transcripts associated with this gene.

View Full Product Details