Product Details

SNP ID
rs12005961
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:135344068 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGCTGACGCCTGGGTCTCCAGA[G/T]ACCCTCCTGGAACTCCGCCAGGCCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C9orf62 PubMed Links

Gene Details

Gene
C9orf62
Gene Name
chromosome 9 open reading frame 62
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173520.2 270 Missense Mutation GAG,GAT E40D NP_775791.1

View Full Product Details