Product Details

SNP ID
rs170296
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41504703 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTGTAGCCTCCCCTCACATGAGGT[G/T]GCTGGAAAGGACAGGATTAGCCTCA
Phenotype
MIM: 147521 MIM: 151520
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ITPKA PubMed Links

Gene Details

Gene
ITPKA
Gene Name
inositol-trisphosphate 3-kinase A
There are no transcripts associated with this gene.

Gene
LTK
Gene Name
leukocyte receptor tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135685.1 Intron NP_001129157.1
NM_002344.5 Intron NP_002335.2
NM_206961.3 Intron NP_996844.1
XM_011521556.2 Intron XP_011519858.1
XM_011521557.2 Intron XP_011519859.1
XM_017022181.1 Intron XP_016877670.1
XM_017022182.1 Intron XP_016877671.1
XM_017022183.1 Intron XP_016877672.1

View Full Product Details