Product Details

SNP ID
rs12706945
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:130511242 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TAAGAGACACTTATGAGGTATTTCC[A/G]GAAAGCTTTGTTAGAATAATAATTA
Phenotype
MIM: 604355 MIM: 601029
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COPG2 PubMed Links

Gene Details

Gene
COPG2
Gene Name
coatomer protein complex subunit gamma 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290033.1 Intron NP_001276962.1
NM_012133.5 Intron NP_036265.3
Gene
MEST
Gene Name
mesoderm specific transcript
There are no transcripts associated with this gene.

View Full Product Details