Product Details

SNP ID
rs35155356
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:86468931 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAAAGAAAGACACCTTCGTAACC[C/T]GCATTTTCCAAAGAGAGAAATCACA
Phenotype
MIM: 603906
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLCA1 PubMed Links

Gene Details

Gene
CLCA1
Gene Name
chloride channel accessory 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001285.3 89 UTR 5 NP_001276.2

View Full Product Details