Product Details

SNP ID
rs9981735
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.21:29195543 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAACAGCAATAATAATAAAAACT[A/G]TGACACCTGAGTCTTATTGATCCCT
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LINC00189 PubMed Links

Gene Details

Gene
LINC00189
Gene Name
long intergenic non-protein coding RNA 189
There are no transcripts associated with this gene.

View Full Product Details