Product Details

SNP ID
rs13251475
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:22988822 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGGGGCTGAGACAGATGCTAGCGAG[A/G]CTATGATAACTCAGACAAGGGGCTG
Phenotype
MIM: 607352
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC107984124 PubMed Links

Gene Details

Gene
LOC107984124
Gene Name
translation initiation factor IF-2-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017014154.1 1417 Intron XP_016869643.1
Gene
RHOBTB2
Gene Name
Rho related BTB domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160036.1 1417 Intron NP_001153508.1
NM_001160037.1 1417 Intron NP_001153509.1
NM_015178.2 1417 Intron NP_055993.2
XM_017013251.1 1417 UTR 5 XP_016868740.1

View Full Product Details