Product Details

SNP ID
rs5930928
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:136308659 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTTGGCTGACATGACTTTAAGCAA[A/T]AGGAAAAAACCCTTAGGTGGTATGC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ADGRG4 PubMed Links

Gene Details

Gene
ADGRG4
Gene Name
adhesion G protein-coupled receptor G4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153834.3 Intron NP_722576.3
XM_011531269.2 Intron XP_011529571.1
XM_011531271.2 Intron XP_011529573.1

View Full Product Details