Product Details

SNP ID
rs1990760
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:162267541 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGTCGGCACACTTCTTTTGCAGTG[C/T]TTTGTTTTCTCTTACAATGTAAAGT
Phenotype
MIM: 606951
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFIH1 PubMed Links

Gene Details

Gene
IFIH1
Gene Name
interferon induced with helicase C domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022168.3 3237 Missense Mutation ACA,GCA T946A NP_071451.2

View Full Product Details