Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001252119.1 | 4092 | Missense Mutation | NP_001239048.1 | ||
NM_001252120.1 | 4092 | Missense Mutation | NP_001239049.1 | ||
NM_001252122.1 | 4092 | Missense Mutation | NP_001239051.1 | ||
NM_001252124.1 | 4092 | Intron | NP_001239053.1 | ||
NM_015148.3 | 4092 | Missense Mutation | NP_055963.2 | ||
XM_005246991.1 | 4092 | Missense Mutation | XP_005247048.1 | ||
XM_011510826.1 | 4092 | Missense Mutation | XP_011509128.1 | ||
XM_011510828.1 | 4092 | Missense Mutation | XP_011509130.1 | ||
XM_011510829.1 | 4092 | Missense Mutation | XP_011509131.1 | ||
XM_011510830.1 | 4092 | Missense Mutation | XP_011509132.1 | ||
XM_011510831.2 | 4092 | Missense Mutation | XP_011509133.1 | ||
XM_011510832.2 | 4092 | Missense Mutation | XP_011509134.1 | ||
XM_011510833.1 | 4092 | Missense Mutation | XP_011509135.1 | ||
XM_011510834.1 | 4092 | Missense Mutation | XP_011509136.1 | ||
XM_011510835.1 | 4092 | Missense Mutation | XP_011509137.1 | ||
XM_017003634.1 | 4092 | Missense Mutation | XP_016859123.1 | ||
XM_017003635.1 | 4092 | Missense Mutation | XP_016859124.1 | ||
XM_017003636.1 | 4092 | Missense Mutation | XP_016859125.1 |