Product Details

SNP ID
rs3773999
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:190516049 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTTATTCTGATGAACTTCATGCAAT[G/A]ATTTACTACTATCTACTGGCTTTGC
Phenotype
MIM: 602626
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
IL1RAP PubMed Links

Gene Details

Gene
IL1RAP
Gene Name
interleukin 1 receptor accessory protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167928.1 Intron NP_001161400.1
NM_001167929.1 Intron NP_001161401.1
NM_001167930.1 Intron NP_001161402.1
NM_001167931.1 Intron NP_001161403.1
NM_002182.3 Intron NP_002173.1
NM_134470.3 Intron NP_608273.1
XM_017006347.1 Intron XP_016861836.1
XM_017006348.1 Intron XP_016861837.1

View Full Product Details