Product Details

SNP ID
rs2241003
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:120904499 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCAATTTGAAGTCCTAGTGGAGCC[C/G]TCTGGGTTTGCTTGTTCACCTCCAA
Phenotype
MIM: 609740 MIM: 601711
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PHF19 PubMed Links

Gene Details

Gene
PHF19
Gene Name
PHD finger protein 19
There are no transcripts associated with this gene.

Gene
TRAF1
Gene Name
TNF receptor associated factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190945.1 2197 UTR 3 NP_001177874.1
NM_001190947.1 2197 UTR 3 NP_001177876.1
NM_005658.4 2197 UTR 3 NP_005649.1

View Full Product Details