Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_014714.3 | 737 | Intron | NP_055529.2 | ||
XM_005255725.4 | 737 | Intron | XP_005255782.1 | ||
XM_005255726.3 | 737 | Intron | XP_005255783.1 | ||
XM_006720990.3 | 737 | Intron | XP_006721053.1 | ||
XM_006720991.3 | 737 | Intron | XP_006721054.1 | ||
XM_006720992.3 | 737 | Intron | XP_006721055.1 | ||
XM_011522766.2 | 737 | Intron | XP_011521068.1 | ||
XM_011522767.2 | 737 | Intron | XP_011521069.1 | ||
XM_011522769.2 | 737 | Intron | XP_011521071.1 | ||
XM_011522771.2 | 737 | Intron | XP_011521073.1 | ||
XM_011522772.2 | 737 | Intron | XP_011521074.1 | ||
XM_017023910.1 | 737 | Intron | XP_016879399.1 | ||
XM_017023911.1 | 737 | Intron | XP_016879400.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
XM_017023923.1 | 737 | Missense Mutation | CCC,TCC | P241S | XP_016879412.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001256541.1 | 737 | Intron | NP_001243470.1 | ||
NM_024600.5 | 737 | Intron | NP_078876.2 |