Product Details

SNP ID
rs2015743
Assay Type
Functionally tested
NCBI dbSNP Submissions
48
Location
Chr.1:210343352 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGCCCCTTCCTTTAAAATAGTTAC[G/T]TTTAAGTTTTCATTTCTGCCTTCGT
Phenotype
MIM: 605743
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HHAT PubMed Links

Gene Details

Gene
HHAT
Gene Name
hedgehog acyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122834.3 489 Intron NP_001116306.1
NM_001170564.2 489 Intron NP_001164035.1
NM_001170580.2 489 Intron NP_001164051.1
NM_001170587.2 489 Intron NP_001164058.1
NM_001170588.2 489 Intron NP_001164059.1
NM_018194.5 489 Intron NP_060664.2
XM_006711441.3 489 Intron XP_006711504.1
XM_011509737.2 489 Intron XP_011508039.1
XM_011509738.2 489 Intron XP_011508040.1
XM_011509739.2 489 Intron XP_011508041.1
XM_011509740.2 489 Intron XP_011508042.1
XM_011509741.2 489 Intron XP_011508043.1
XM_011509742.2 489 Intron XP_011508044.1
XM_011509744.2 489 Intron XP_011508046.1
XM_011509746.2 489 Intron XP_011508048.1
XM_011509747.1 489 Intron XP_011508049.1
XM_017001726.1 489 Intron XP_016857215.1
XM_017001727.1 489 Intron XP_016857216.1
XM_017001728.1 489 Intron XP_016857217.1
XM_017001729.1 489 Intron XP_016857218.1
XM_017001730.1 489 Intron XP_016857219.1
XM_017001731.1 489 Intron XP_016857220.1
XM_017001732.1 489 Intron XP_016857221.1
XM_017001733.1 489 Intron XP_016857222.1
XM_017001734.1 489 Intron XP_016857223.1
XM_017001735.1 489 UTR 5 XP_016857224.1
XM_017001736.1 489 Intron XP_016857225.1
XM_017001737.1 489 Intron XP_016857226.1
XM_017001738.1 489 Intron XP_016857227.1
XM_017001739.1 489 Intron XP_016857228.1
XM_017001740.1 489 Intron XP_016857229.1
XM_017001741.1 489 Intron XP_016857230.1
XM_017001742.1 489 Intron XP_016857231.1
XM_017001743.1 489 Intron XP_016857232.1
XM_017001744.1 489 Intron XP_016857233.1

View Full Product Details