Product Details

SNP ID
rs2075436
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:109280514 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAGGCTGTGGCACGGGTTAAACAA[C/G]AGGACTTGTGAAAAGCATTTAGTTC
Phenotype
MIM: 609429
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FOXN4 PubMed Links

Gene Details

Gene
FOXN4
Gene Name
forkhead box N4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_213596.2 Intron NP_998761.2
XM_011537922.2 Intron XP_011536224.1
XM_011537923.2 Intron XP_011536225.1
XM_017018818.1 Intron XP_016874307.1
XM_017018819.1 Intron XP_016874308.1

View Full Product Details