Product Details

SNP ID
rs9819960
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:128065670 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTGCTGAGACTAAGCCACATTC[C/T]TGCTTAATAAAAAGCTATCTCTTTA
Phenotype
MIM: 603449 MIM: 609213
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RUVBL1 PubMed Links

Gene Details

Gene
RUVBL1
Gene Name
RuvB like AAA ATPase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319084.1 Intron NP_001306013.1
NM_001319086.1 Intron NP_001306015.1
NM_003707.2 Intron NP_003698.1
XM_005247841.3 Intron XP_005247898.1
XM_011513248.1 Intron XP_011511550.1
XM_011513249.2 Intron XP_011511551.1
XM_017007356.1 Intron XP_016862845.1
XM_017007357.1 Intron XP_016862846.1
Gene
SEC61A1
Gene Name
Sec61 translocon alpha 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013336.3 Intron NP_037468.1

View Full Product Details