Product Details

SNP ID
rs2231675
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:99732484 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGGCCGGCGGGAGTCGTAGTCAGT[A/G]GTGATTTCTTCTGGAGTCGTTTCCT
Phenotype
MIM: 603646 MIM: 610101
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COX15 PubMed Links

Gene Details

Gene
COX15
Gene Name
COX15, cytochrome c oxidase assembly homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320974.1 183 UTR 5 NP_001307903.1
NM_001320975.1 183 UTR 5 NP_001307904.1
NM_001320976.1 183 UTR 5 NP_001307905.1
NM_004376.6 183 UTR 5 NP_004367.2
NM_078470.5 183 UTR 5 NP_510870.1
XM_006717634.3 183 Intron XP_006717697.1
XM_011539298.2 183 Intron XP_011537600.1
XM_017015715.1 183 UTR 5 XP_016871204.1
Gene
CUTC
Gene Name
cutC copper transporter
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015960.2 183 Intron NP_057044.2

View Full Product Details