Product Details

SNP ID
rs8100458
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40994308 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAACAGAGTAAGCAAACCTCAAGA[C/T]ACTCAAGGTAGGCACTCGTGGTGTT
Phenotype
MIM: 123930
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYP2B6 PubMed Links

Gene Details

Gene
CYP2B6
Gene Name
cytochrome P450 family 2 subfamily B member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000767.4 Intron NP_000758.1
XM_005258569.4 Intron XP_005258626.1
XM_006723050.3 Intron XP_006723113.1
XM_011526546.2 Intron XP_011524848.1
XM_011526547.2 Intron XP_011524849.1
XM_011526548.2 Intron XP_011524850.1
XM_011526549.2 Intron XP_011524851.1
XM_011526550.2 Intron XP_011524852.1

View Full Product Details