Product Details

SNP ID
rs2438514
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36215731 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGGTGCCTACTTCAGACTATTCAA[A/G]TCCCCATCTTTGGGGTACTTAAATA
Phenotype
MIM: 601505 MIM: 614275
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF146 PubMed Links

Gene Details

Gene
ZNF146
Gene Name
zinc finger protein 146
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099638.1 Intron NP_001093108.1
NM_001099639.1 Intron NP_001093109.1
NM_007145.2 Intron NP_009076.2
XM_005259214.2 Intron XP_005259271.1
XM_017027244.1 Intron XP_016882733.1
XM_017027245.1 Intron XP_016882734.1
XM_017027246.1 Intron XP_016882735.1
XM_017027247.1 Intron XP_016882736.1
Gene
ZNF565
Gene Name
zinc finger protein 565
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042474.2 Intron NP_001035939.1
NM_152477.4 Intron NP_689690.3
XM_011526512.2 Intron XP_011524814.1
XM_011526514.2 Intron XP_011524816.1
XM_017026341.1 Intron XP_016881830.1
XM_017026342.1 Intron XP_016881831.1
XM_017026343.1 Intron XP_016881832.1

View Full Product Details