Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001040458.1 | Intron | NP_001035548.1 | |||
NM_001198541.1 | Intron | NP_001185470.1 | |||
NM_016442.3 | Intron | NP_057526.3 | |||
XM_005272015.4 | Intron | XP_005272072.1 | |||
XM_005272016.4 | Intron | XP_005272073.1 | |||
XM_011543480.2 | Intron | XP_011541782.1 | |||
XM_011543481.2 | Intron | XP_011541783.1 | |||
XM_011543484.2 | Intron | XP_011541786.1 | |||
XM_011543485.2 | Intron | XP_011541787.1 | |||
XM_011543486.2 | Intron | XP_011541788.1 | |||
XM_017009580.1 | Intron | XP_016865069.1 | |||
XM_017009581.1 | Intron | XP_016865070.1 | |||
XM_017009582.1 | Intron | XP_016865071.1 | |||
XM_017009583.1 | Intron | XP_016865072.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
XM_011543544.1 | Intron | XP_011541846.1 |