Product Details

SNP ID
rs12447568
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:75445018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATTACATCCAAAGAATGCTTTA[A/G]GCAAGAACTCCTTCAAAAGACATTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM170A PubMed Links

Gene Details

Gene
TMEM170A
Gene Name
transmembrane protein 170A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304996.1 3369 UTR 3 NP_001291925.1
NM_001304997.1 3369 UTR 3 NP_001291926.1
NM_001304998.1 3369 UTR 3 NP_001291927.1
NM_145254.2 3369 UTR 3 NP_660297.1
XM_011522869.2 3369 UTR 3 XP_011521171.1
XM_017022939.1 3369 UTR 3 XP_016878428.1
XM_017022940.1 3369 UTR 3 XP_016878429.1
XM_017022941.1 3369 UTR 3 XP_016878430.1
XM_017022942.1 3369 Intron XP_016878431.1
XM_017022943.1 3369 UTR 3 XP_016878432.1

View Full Product Details