Product Details

SNP ID
rs11654535
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39463394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAACGTCAAAGGCTTCTGCTTA[G/T]TGAGTGGATGGAGTTGATACTAGTT
Phenotype
MIM: 615514
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CDK12 PubMed Links

Gene Details

Gene
CDK12
Gene Name
cyclin dependent kinase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015083.2 Intron NP_055898.1
NM_016507.3 Intron NP_057591.2
XM_005257456.3 Intron XP_005257513.1
XM_005257458.4 Intron XP_005257515.1
XM_011524892.2 Intron XP_011523194.1
XM_011524893.2 Intron XP_011523195.1
XM_011524894.2 Intron XP_011523196.1
XM_011524895.2 Intron XP_011523197.1
XM_011524896.2 Intron XP_011523198.1
XM_011524897.2 Intron XP_011523199.1
XM_011524898.2 Intron XP_011523200.1
XM_011524899.2 Intron XP_011523201.1
XM_011524900.2 Intron XP_011523202.1
XM_011524901.2 Intron XP_011523203.1
XM_011524902.2 Intron XP_011523204.1
XM_011524903.2 Intron XP_011523205.1
XM_011524905.2 Intron XP_011523207.1
XM_011524906.2 Intron XP_011523208.1
XM_011524907.2 Intron XP_011523209.1
XM_017024744.1 Intron XP_016880233.1
XM_017024745.1 Intron XP_016880234.1
XM_017024746.1 Intron XP_016880235.1
XM_017024747.1 Intron XP_016880236.1
XM_017024748.1 Intron XP_016880237.1
XM_017024749.1 Intron XP_016880238.1
XM_017024750.1 Intron XP_016880239.1
XM_017024751.1 Intron XP_016880240.1
XM_017024752.1 Intron XP_016880241.1
XM_017024753.1 Intron XP_016880242.1

View Full Product Details