Product Details
- SNP ID
-
rs9926
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:10075236 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACGTGTACAGACACCCTGCAGAGAC[A/G]AGGGTCATGACTGGGCCCTCCTGCC
- Phenotype
-
MIM: 604734
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR3138
PubMed Links
Gene Details
- Gene
- MIR3138
- Gene Name
- microRNA 3138
There are no transcripts associated with this gene.
- Gene
- WDR1
- Gene Name
- WD repeat domain 1
View Full Product Details