Product Details

SNP ID
rs12985721
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40934109 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGCATTTGTAAGTAAACTTAAAC[A/G]TTTTTCTAATTTTTATGTCAAGTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYP2B7P PubMed Links

Gene Details

Gene
CYP2B7P
Gene Name
cytochrome P450 family 2 subfamily B member 7, pseudogene
There are no transcripts associated with this gene.

View Full Product Details