Product Details

SNP ID
rs12971648
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41024773 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTTCAACTGTTTAGCTACTCAGA[C/T]GGGGCTGGTATGAGGGGAGTGTCAC
Phenotype
MIM: 123930
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYP2A7P1 PubMed Links

Gene Details

Gene
CYP2A7P1
Gene Name
cytochrome P450 family 2 subfamily A member 7 pseudogene 1
There are no transcripts associated with this gene.

Gene
CYP2B6
Gene Name
cytochrome P450 family 2 subfamily B member 6
There are no transcripts associated with this gene.

View Full Product Details