Product Details

SNP ID
rs11996576
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:41645484 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATCCTCCACCCAGCCGGAGGGGCT[C/G]TCCTTCTCCCTTCCTGCCTTGTTGG
Phenotype
MIM: 612641 MIM: 610772
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
ANK1 PubMed Links

Gene Details

Gene
ANK1
Gene Name
ankyrin 1
There are no transcripts associated with this gene.

Gene
NKX6-3
Gene Name
NK6 homeobox 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152568.3 1377 UTR 3 NP_689781.1
XM_017013143.1 1377 UTR 3 XP_016868632.1

View Full Product Details