Product Details
- SNP ID
-
rs2570233
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:51829649 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCACGGGAGTGCGCACGCGCGTCGG[A/G]GGCGAAGGAGGAACCCACCGCACCT
- Phenotype
-
MIM: 605112
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TMOD3
PubMed Links
Gene Details
- Gene
- TMOD3
- Gene Name
- tropomodulin 3
View Full Product Details