Product Details

SNP ID
rs279939
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:103048516 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGACCTAACATTTACATTAGTTTT[C/G]TTGCGTGCCTTCACTTTATATTCCT
Phenotype
MIM: 601295
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLC10A2 PubMed Links

Gene Details

Gene
SLC10A2
Gene Name
solute carrier family 10 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000452.2 Intron NP_000443.1

View Full Product Details