Product Details

SNP ID
rs34504992
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:125446177 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GTTGCAGGTGGGGACCTCCGAGCCC[A/T]GGATAGCCCTAGCTGAGAGCAGTCA
Phenotype
MIM: 604825
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FEZ1 PubMed Links

Gene Details

Gene
FEZ1
Gene Name
fasciculation and elongation protein zeta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005103.4 Intron NP_005094.1
NM_022549.3 Intron NP_072043.1
XM_005271734.2 Intron XP_005271791.1
XM_005271735.2 Intron XP_005271792.1

View Full Product Details