Product Details
- SNP ID
-
rs11070641
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:48408832 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCCACTGTGTGCCAACTCCATTTTC[C/T]CCTGTTGTTTGTTATAAAGAAAATC
- Phenotype
-
MIM: 134797
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FBN1
PubMed Links
Gene Details
- Gene
- FBN1
- Gene Name
- fibrillin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000138.4 |
11169 |
UTR 3 |
|
|
NP_000129.3 |
View Full Product Details